Learn More
Cytiva Accessories for Multiphor™ II Electrophoresis System
2255.00 DKK - 5585.00 DKK
Specifications
For Use With (Equipment) Multiphor II Electrophoresis System
View More Specs
Products 8
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10299244
View Documents
Cytiva
18-1004-40
IEF Electrode Strip
2255.00 DKK
Pack of 100
In Stock
to see stock.
Add to basket
10006184
View accessory products View Documents
Cytiva
18-1013-73
Flexiclamp N/A N/A N/A
10371475
View Documents
Cytiva
18-1122-19
EPH Electrode Cathode N/A N/A N/A
10331525
View Documents
Cytiva
18-1122-20
EPH Electrode Anode N/A N/A N/A
11590824
View Documents
Cytiva
1850-913
EPH Electrode Wick (82 x 130 mm) N/A N/A N/A
10127294
View Documents
Cytiva
80112953
EPH Electrode Wick (82 x 130 mm) N/A N/A N/A
11580824
View Documents
Cytiva
1850-912
EPH Electrode Wick (104 x 253 mm) N/A N/A N/A
10586555
View Documents
Cytiva
80112952
EPH Electrode Wick (104 x 253 mm) N/A N/A N/A
Specifications
Multiphor II Electrophoresis System
Documents
Product Certifications
Product Content Correction
Product Title
Cytiva Accessories for Multiphor™ II Electrophoresis System