Learn More
Cytiva Accessories for SE 250/260 Mini-vertical Gel Units: Alumina Notched Plate
Brand:  Cytiva SE202N
486.43 DKK valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 10279714
  • 565.00 DKK / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Alumina
8 cm
Notched Alumina Plate
10 cm