Learn More
Micronova™ Accordion Over-Under™ Handle
Designed to deal with the unique space limitations in isolators and containment hoods
Brand:  Micronova™ QDAH-08
Product Code. 15992221
  • 1445.00 DKK / Each
Only null left
Estimated Shipment: 10-10-2025
Description
  • Each section features a polypropylene hinge and quick disconnect to allow for attachment of additional sections and ultimately the mop adapter
  • The unique design allows the operator to fit the collapsed handle into the pass thru and once inside the isolator provides the extended reach necessary to cover all surfaces
  • Outside of the Isolator, when connected to a standard handle, the Accordion Handle allows the operator to angle the mop head to effectively and comfortably clean ‘over and under’ obstacles in a cleanroom or controlled environment
  • Autoclavable
Specifications
Specifications
Sterile
8 in.
Stainless steel handle with locking swivel action that holds various adapters and works as an extension to reach over and under equipment
Electropolished Stainless Steel (304), Polyethylene
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number047603-000 Certificate TypeCertificate of Compliance Date16/01/2025 Catalog NumberQDAH-08
Lot Number047482-000 Certificate TypeCertificate of Compliance Date31/12/2024 Catalog NumberQDAH-08
Lot Number047430-000 Certificate TypeCertificate of Compliance Date31/12/2024 Catalog NumberQDAH-08
Lot Number046283-000 Certificate TypeCertificate of Compliance Date11/12/2024 Catalog NumberQDAH-08
Lot Number041890-000 Certificate TypeCertificate of Compliance Date23/01/2024 Catalog NumberQDAH-08
Promotions
Promotions
Product Content Correction
Product Title
Micronova™ Accordion Over-Under™ Handle > Width: 203.2 (8 in.)