Learn More
Brand™ accu-jet™ pro Pipet Controller
Provides fatigue-free pipetting even in prolonged operations with approximately eight hours of continuous pipetting without recharging. BRAND™ accu-jet™ pro Pipet Controller is designed for one-handed delivery mode and speed selection. A 0.2μm hydrophobic membrane filter and a built-in check valve protect against liquid penetrating inside.
Brand:  Brand™ 26303
 View more versions of this product
Product Code. 10081422
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • One-handed delivery mode and selection
  • Ergonomic design is perfectly balanced, weighing only 190g for fatigue-free pipetting even in prolonged operations.
  • Adjustable motor speed limiter – a slower speed setting allows even more sensitive control of filling and dispensing rates for lower volume pipets.
  • A flashing LED indicates 2 hours in advance when the battery needs charging. Approximately eight hours of continuous pipetting (with a 10mL pipet) without recharging.
  • Direct outlet of liquid vapors protects against corrosion to ensure long instrument life.
  • A 0.2ÎĽm hydrophobic membrane filter and a built-in check valve protect against liquid penetrating inside. Perfect fit for glass and plastic pipets from 0.1 to 200mL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nickel-metal hydride
0.2 ÎĽm
Hydrophobic membrane
Europe (continent)
190 g
50/60 Hz
Royal Blue
Pipette Controller
Nickel-metal hydride battery, 2 battery compartment covers, wall support, AC adapter (100-240V; 50/60Hz) and 2 spare membrane filters 0.2µm, sterile. 26304 without recharger
1 Each
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brand™ accu-jet™ pro Pipet Controller > Royal blue