missing translation for 'onlineSavingsMsg'
Learn More
Acetic acid, 99.7%, Scharlab Product Code.: 11312655
compliance-icons compliance-icons

Acetic acid, 99.7%, Scharlab

Product Code. 11312655
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11312655

Brand: Scharlab AC03421000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 64-19-7
Molecular Formula C2H4O2
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
Grade Pharmpur™
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • Acetic acid glacial, extra pure, Pharmpur, Ph Eur, BP, USP
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic acid, 99.7%, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.