missing translation for 'onlineSavingsMsg'
Learn More
Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™ Product Code.: 15621600

Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15621600
1 L, Glass Bottle, 1 L
Click to view available options
Quantity:
500 mL
1 L
2 L
2.5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
2 L
2.50 L
20 L
500mL
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15621600

Brand: Honeywell-Riedel-de Haen 1791241L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetoneShow MoreShow Less
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Boiling Point 55°C to 57°C
Quantity 1 L
Flash Point −17°C
Packaging Glass Bottle
Linear Formula CH3COCH3
UN Number UN1090
Beilstein 635680
Formula Weight 58.08
Percent Purity >99.5%
Grade ACS Reagent
Assay acc. to ACS
Chemical Name or Material Acetone, ACS reagent, ≥99.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetone, ACS reagent, ≥99.5%, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.