missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, for HPLC, Fisher Chemical™

Product Code. 11473493 Shop All Fisher Chemical Products
2.5 L, Amber Glass Bottle, 2.5 L
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Unit Size:
1 L
2.5 L
5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

C2H3N, CAS Number-75-05-8, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053, Amber glass bottle, Liquid, 6342, CC#N, 1.42, 97mbar at 20 deg.C, 0.36cp at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 81°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C2H3N
Vapor Pressure 97 mbar at 20°C
Viscosity 0.36 cp at 20°C
Molecular Weight (g/mol) 41.05
Formula Weight 41.05 g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, for HPLC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.