missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11594-1-AP-150UL
This item is not returnable.
View return policy
Description
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher′s disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin′s lymphoma and in the sera of patients undergoing active bone turnover.Specifications
ACP5 | |
Polyclonal | |
Unconjugated | |
ACP5 | |
ACP5, Tartrate resistant acid ATPase, TR AP, TRACP, TRAP, TrATPase, Type 5 acid phosphatase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11433, 25732, 54 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13686, P29288, Q05117 | |
ACP5 | |
ACP5 Fusion Protein Ag2065 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction