Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance Multi-well Plate Lids
Multi-well plate lids not approved for use in medical, clinical, surgical or other patient applications.
Brand:  Cytiva (formerly Pall Lab) 8001
Product Code. 17104601
  • 565.00 DKK / Pack of 10
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Prefilter provides efficient clarification of highly particulate samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Non-sterile
10/Pk.