Learn More
Thermo Scientific Chemicals Glass beads, 400-800 micron, Thermo Scientific Chemicals
Brand: Thermo Scientific Chemicals 397645000
1462.35 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glass | |
500 g |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0463208 | Certificate of Analysis | 18/06/2024 | 397640250,397641000,397645000 |
A0458519 | Certificate of Analysis | 06/12/2023 | 397640250,397641000,397645000 |
A0450372 | Certificate of Analysis | 02/05/2023 | 397640250,397641000,397645000 |
A0404490 | Certificate of Analysis | 13/12/2022 | 397640250,397641000,397645000 |
A0416167 | Certificate of Analysis | 12/12/2022 | 397640250,397641000,397645000 |