Learn More
Thermo Scientific Chemicals Glass Wool Product Code.: 11927001
Thermo Scientific Chemicals Glass Wool
Product Code. 11927001
1kg
Click to view available options
Quantity:
50g
250g
1kg
Product Code. 11927001
Brand: Thermo Scientific Chemicals 386060010
Request Bulk or Custom Format
Filter aid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1kg
Boiling Point 1000°C
Product Type Glass Wool
For Use With (Application) Filter aid
Material Glass
Melting Point 680°C
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464101 Certificate TypeCertificate of Analysis Date26/06/2024 Catalog Number386060500,386062500,386060010
Lot NumberA0459048 Certificate TypeCertificate of Analysis Date03/01/2024 Catalog Number386060500,386062500,386060010
Lot NumberA0448814 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number386060500,386062500,386060010
Lot NumberA0453702 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number386060500,386062500,386060010
Lot NumberA0397990 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number386060500,386062500,386060010
compliance-icons
Product Identifier
  • Glass wool
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155
    Expires2025-04-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Glass Wool >