Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Adapter for Rotors designed for use with Eppendorf Centrifuges
Brand:  Eppendorf™ 5427736000
640.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10676654
  • 642.56 DKK / Pack of 2
Description
  • Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Large Rotor Bore Adapter
1 x 30 mL Tube
Rotor F-35-6-30
26 dia. x 92 to 112mm
Round Bottom
26 mm
Large rotor bore
2/Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Adapter for Rotor F-35-6-30 > Device Holds: 2.6 to 7mL; Tube Type: Round bottom tube