Learn More
Eppendorf™ Adapter for Rotor F-45-64-5-PCR
For use with PCR tube rotor
Brand: Eppendorf™ 5427717005
1470.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adapter for Rotor F-45-64-5-PCR used with Eppendorf Centrifuge
- Rotor F-45-64-5-PCR holds up to 64 × 0.2mL PCR tubes
Specifications
Large Rotor Bore Adapter | |
F-45-64-5PCR | |
PCR Strips |
PCR | |
Adapter | |
4/Pk. |
Product Content Correction
Product Title
Eppendorf™ Adapter for Rotor F-45-64-5-PCR > Device Holds: PCR Strips; Tube Type: PCR
Spot an opportunity for improvement?Share a Content Correction