Learn More
BRAND™ Polypropylene Adapter for Pipette Tips
Polypropylene construction. For use with 100μL pipette tips.
Brand: BRAND™ 25961
54.83 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Polypropylene construction
- For 100μL pipette tips
Specifications
Polypropylene | |
Tip Adapter | |
100μL pipette tips |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
16Hours
23Minutes
45Seconds
6New Offer Save Up to 92%