Learn More
Eppendorf™ Adapters For Fixed-Angle Conical Falcon™ Rotor
Use these adaptors with Fixed-Angle Conical Falcon™ Rotors.
Brand: Eppendorf™ 5804776003
720.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Set of 2 adaptors
- Compatible with Rotor F-34-6-38
Specifications
Adapter | |
15 mL | |
17 mm | |
2 Pack |
Conical Falcon | |
Fixed angle rotor | |
6 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
Eppendorf™ Adapters For Fixed-Angle Conical Falcon™ Rotor > Capacity: 15mL; Number of Places: 6
Spot an opportunity for improvement?Share a Content Correction