Learn More
Memmert™ Additional Stainless Steel Grid for SN260, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ SN260E29766
This item is not returnable.
View return policy
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
SN260 Sterilizer | |
Reinforced Grid | |
Stainless Steel |