Learn More
3M™ Adflo™ Lithium Ion Battery
3M™ Adflo™ Lithium Ion Battery makes the Adflo™ Powered Air Respirator even more lightweight and balanced for greater comfort during the full shift.
Brand: 3M™ 7100166697
5909.96 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- With the right type of filter, the Adflo respirator effectively protects you against both particles and gases - all in one system. Gas filters are sold as separate accessories
- Incorporating a Li-ion battery, making the turbo almost 20% lighter in weight
- To remove unpleasant smells an odor filter can be added (not needed if a gas filter is installed) - sold separately
- The airflow is always a constant nominal rate of 170 L per minute, regardless of the battery's charge or the particle loading of the filter
- For hot, humid jobs, you can increase the airflow to 200 L per minute by simply pushing the ON button a second time
- The lightweight Lithium-ion battery fully charges in 4.5 hours and has a operating time of 7-9 hours. If you like to extend operating time, there is a heavy-duty battery option available
- To order a complete welding helmet system, first select headtop
Specifications
7 to 9 hr. | |
EN 12941 | |
Particulate HE Filter |
Lithium-ion | |
PARP Air Source |