missing translation for 'onlineSavingsMsg'
Learn More

Advanced Instruments 621 Cryoscope Standard Bottle

Standard bottle used when calibrating and checking the performance of cryoscopes

Brand:  Advanced Instruments 3LA032

Encompass

Product Code. 15715682

  • 422.76 DKK / 110mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Capacity: 110mL (3.7 oz)
  • Expected Value: 621 m°H (-600 m°C)
  • Expected Range: 619 - 623 m°H (-598 to -602 m°C)

Specifications

Specifications

Calibration Standard Bottle
Cryoscope, osmometers and other calibration equipment
110mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Advanced Instruments 621 Cryoscope Standard Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.