Learn More
Motic™ AE31E Binocular Microscope
Use this microscope for routine live cell inspection. Motic™ AE31E Inverted Binocular Microscope is ideal for university, routine-clinical and lab environments.
Brand: Motic™ 1100100200565
This item is not returnable.
View return policy
Description
- Centering telescope.
- CCIS Plan achromatic objective PL4X
- CCIS Plan achromatic Phase objective PL PH10X, LWD PL PH20X, LWD PL PH40X
- Plain stage, glass and metal inserts
- Phase slider, centerable PH1, brightfield, PH3
Specifications
100/240 V | |
Side-facing quintuple nosepiece. | |
Widefield High Eyepoint WFPL10X/22 mm with diopter adjustment on both eyepieces and rubber eyecups | |
Inverted Binocular Microscope |
ELWD N.A. 0.3 with phase slider (Ph1, BF) | |
Universal power supply 100/240 V (CE) | |
Quartz Halogen |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Motic™ AE31E Binocular Microscope > Product Type: Inverted Binocular Microscope
Spot an opportunity for improvement?Share a Content Correction