Learn More
Agar-Agar, Pure (Granular Powder), Fisher Chemical™
Brand: Fisher Chemical A/1080/53
1433.22 DKK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Specifications
Beige | |
Plastic Powder Jar | |
500 g |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
67Hours
11Minutes
15Seconds
52New Offer Save Up to 59% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
12Hours
11Minutes
15Seconds
52