Learn More
Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 2mm, I.D.: 1mm, Thermo Scientific Chemicals
156.00 DKK - 470.89 DKK
Specifications
Chemical Name or Material | Al-23 Insulating Tube |
---|---|
Name Note | Both ends open (Thin Wall) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367508
|
Thermo Scientific Alfa Aesar
031487.DM |
100 mm | N/A | ||||||
11387508
|
Thermo Scientific Alfa Aesar
031487.DR |
200 mm |
321.68 DKK
200mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377508
|
Thermo Scientific Alfa Aesar
031487.DW |
300 mm |
470.89 DKK
300mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Al-23 Insulating Tube | |
Both ends open (Thin Wall) |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 2mm, I.D.: 1mm, Thermo Scientific Chemicals