Learn More
Al-23 Solid Rod, Round, Dia.: 3mm, Thermo Scientific™
143.00 DKK - 773.51 DKK
Specifications
Chemical Name or Material Al-23 Solid Rod
Form Rod
View More Specs
Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373058
View Documents
Thermo Scientific Alfa Aesar
032927.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11333068
View Documents
Thermo Scientific Alfa Aesar
032927.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11383058
View Documents
Thermo Scientific Alfa Aesar
032927.DT
250 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11343068
View Documents
Thermo Scientific Alfa Aesar
032927.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11393058
View Documents
Thermo Scientific Alfa Aesar
032927.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11313068
View Documents
Thermo Scientific Alfa Aesar
032927.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11323068
View Documents
Thermo Scientific Alfa Aesar
032927.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Solid Rod
Rod
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Al-23 Solid Rod, Round, Dia.: 3mm, Thermo Scientific™