Learn More
Al-23 Tube, Both Ends Open, O.D.: 24mm, I.D.: 18mm, Thermo Scientific Chemicals
409.07 DKK - 2071.00 DKK
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353997
View Documents
Thermo Scientific Alfa Aesar
031572.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11303997
View Documents
Thermo Scientific Alfa Aesar
031572.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11333997
View Documents
Thermo Scientific Alfa Aesar
031572.DW
300 mm
1559.64 DKK
300mm
11343997
View Documents
Thermo Scientific Alfa Aesar
031572.EH
600 mm
1784.59 DKK
600mm
11323997
View Documents
Thermo Scientific Alfa Aesar
031572.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11393987
View Documents
Thermo Scientific Alfa Aesar
031572.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures