Learn More
Al-23 Tube, Both Ends Open, O.D.: 25mm, I.D.: 20mm, Thermo Scientific Chemicals
377.00 DKK - 2864.00 DKK
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324007
View Documents
Thermo Scientific Alfa Aesar
031573.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11304007
View Documents
Thermo Scientific Alfa Aesar
031573.DR
200 mm
802.31 DKK
200mm
11383997
View Documents
Thermo Scientific Alfa Aesar
031573.DW
300 mm
937.28 DKK
300mm
11363997
View Documents
Thermo Scientific Alfa Aesar
031573.EH
600 mm
1859.57 DKK
600mm
11393997
View Documents
Thermo Scientific Alfa Aesar
031573.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373997
View Documents
Thermo Scientific Alfa Aesar
031573.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures