Learn More
BINDER™ Alarm Output
Alarm output, zero-voltage, accessible via 3-pin clamping sleeve
Brand:  BINDER™ 8012-2503
 View more versions of this product
Product Code. 30238346
  • 3304.02 DKK / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Alarm output
  • Zero-voltage
  • Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
  • Acoustic signal that can be switched off
Specifications
Specifications
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Copper
Analog Output
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Alarm Output > For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600