Learn More
BINDER™ Alarm Output
Alarm output, zero-voltage, accessible via 3-pin clamping sleeve
Brand:  BINDER™ 8012-2504
 View more versions of this product
Product Code. 30238352
  • 3304.02 DKK / Each
Estimated Shipment: 06-06-2025
Description
  • Alarm output
  • Zero-voltage
  • Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
  • Acoustic signal that can be switched off
Specifications
Specifications
KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600
Copper
Analog Output
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Alarm Output > For use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600