Learn More
BINDER™ Alarm Output
Alarm output, zero-voltage, accessible via 3-pin clamping sleeve
Brand: BINDER™ 8012-2370
This item is not returnable.
View return policy
Description
- Alarm output
- Zero-voltage
- Accessible via 3-pin clamping sleeve (max. 24V/2.5A)
- Acoustic signal that can be switched off
Specifications
KB 65/130/260/470/720/1060 | |
Aluminum | |
Analog Output |
Product Content Correction
Product Title
BINDER™ Alarm Output > For use with KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060
Spot an opportunity for improvement?Share a Content Correction