missing translation for 'onlineSavingsMsg'
Learn More
Molecular Probes™ Alexa Fluor™ 610-X NHS Ester (Succinimidyl Ester) Product Code.: 10358553

Molecular Probes™ Alexa Fluor™ 610-X NHS Ester (Succinimidyl Ester)

Product Code. 10358553
1 mg, Each
Click to view available options
Quantity:
1 mg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10358553

Brand: Molecular Probes™ A30050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 610-X dye is water soluble and pH-insensitive from pH 4 to pH 10.

Alexa Fluor™ 610-X is a bright and photostable Texas Red™ dye substitute. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 610-X dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 610–R-Phycoerythrin tandem antibody conjugates for flow cytometry.The NHS ester (or succinimidyl ester) of Alexa Fluor™ 610-X is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:

Fluorophore label: Alexa Fluor™ 610-X dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 603/623 nm
Extinction coefficient: 144,000 cm-1M-1
Spectrally similar dyes: Texas Red™
Molecular weight: 1284.8

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 ÎĽg (A33087)
Antibody Conjugate Purification kit for 50-100 ÎĽg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Dye Substitute
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Alexa Fluor 610
Label Type Alexa Fluor
Product Line Alexa Fluor™
Emission 623 nm
Excitation 603 nm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number2151769 Certificate TypeCertificate of Analysis Date09/10/2019 Product CodeA30050
Lot Number1186254 Certificate TypeCertificate of Analysis Date13/04/2017 Product CodeA30050
Lot Number2806830 Certificate TypeCertificate of Analysis Date04/12/2023 Product CodeA30050
Lot Number2486553 Certificate TypeCertificate of Analysis Date28/04/2022 Product CodeA30050
Lot Number2448079 Certificate TypeCertificate of Analysis Date21/12/2021 Product CodeA30050

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Alexa Fluor™ 610-X NHS Ester (Succinimidyl Ester) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.