missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH4 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-06195AF405
This item is not returnable.
View return policy
Description
ALKBH4 Polyclonal specifically detects ALKBH4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ALKBH4 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ABH4, ALKBH4 alkB, alkylation repair homolog 4 (E. coli) | |
Produced in rabbits immunized with E. coli-derived Human ALKBH4 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54784 | |
Store at 4°C in the dark. | |
IgG |