Learn More
alpha-Terpinene, 90+%, Thermo Scientific Chemicals
Description
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.
Solubility
Soluble in ether, alcohol, and ethanol (100 mg/ml). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: -20°C. Store under inert gas. Air sensitive. Keep away from strong acids, strong bases.

Specifications
Specifications
Color | Colorless |
Density | 0.837 |
Boiling Point | 173°C to 175°C |
Flash Point | 50°C (122°F) |
Refractive Index | 1.478 |
Quantity | 500 mL |
UN Number | UN2319 |
Beilstein | 1853379 |
Merck Index | 14,9170 |
Solubility Information | Soluble in ether,alcohol,and ethanol (100mg/ml). Insoluble in water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S02J056 | Certificate of Analysis | 13/06/2023 | J62871.AP |
Q28I058 | Certificate of Analysis | 17/11/2022 | J62871.AE,J62871.AP |
N23B030 | Certificate of Analysis | 07/12/2021 | J62871.AE,J62871.AP |
Z01D069 | Certificate of Analysis | 06/12/2021 | J62871.AE,J62871.AP |
G26Z005 | Certificate of Analysis | 06/12/2021 | J62871.AE,J62871.AP |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.