Learn More
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
- Filter 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low linting
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | X-Large |
Gender | Unisex |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Closure Type | Zipper |
Disposable | Reusable |
Resistance | Fluids |
Style | Knee-length |
Model | 2000 |
Show More |
Product Content Correction
Product Title
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound >
Spot an opportunity for improvement?Share a Content Correction