Learn More
Thermo Scientific™ ALPS Heat Sealer Accessories
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Brand: Thermo Scientific™ AB-0563/1000
1826.21 DKK valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
Warranty: 1 year
Specifications
96-Well PCR Plate Carrier | |
ALPS™ Heat Sealer | |
1 Ea. | |
Carrier |
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30