Learn More
Bochem™ Aluminum Lab Jacks
1405.00 DKK - 4830.00 DKK
Specifications
Material | Aluminum, Stainless Steel |
---|---|
Product Type | Lab Jack |
Quantity | 1 Ea. |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | ||||||
10313712
|
Bochem™
11022 |
130 mm | N/A | |||||||
10350341
|
Bochem™
11015 |
100 mm | N/A | |||||||
10211542
|
Bochem™
11032 |
200 mm |
1730.00 DKK
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
10410251
|
Bochem™
11080 |
300 mm |
4830.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Specifications
Aluminum, Stainless Steel | |
Lab Jack | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bochem™ Aluminum Lab Jacks