Learn More
Bochem™ Aluminum Rod for Stand Base M10
The aluminum rods with M10 thread for used with Bochem stand bases and clamps. For holding and supporting laboratory apparatus.
Brand:  Bochem™ 5133
Product Code. 10320631
  • 151.00 DKK / Each
Only null left
Estimated Shipment: 01-08-2025
Description
  • Aluminum
  • Thread M10
Specifications
Specifications
Aluminum
12 mm
Holdaing and Supporting Laboratory Apparatus
1000 mm
Aluminum Stand Rod M10
Stand Base M10, Clamps
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Aluminum Rod for Stand Base M10 > Aluminum Rods; Length: 1000 mm