Learn More
Fisherbrand™ Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Bossheads come with offset jaws for greater strength.
Brand:  Fisherbrand™ 9507/P
 View more versions of this product
Product Code. 11547722
  • 440.00 DKK / Pack of 5
Estimated Shipment: 26-05-2025
Explore more special offers
Add to basket
Description
Description
  • Aluminum alloy with clamping screws
  • For use with rods up to 16mm in diameter
  • Screws have plastic heads
Specifications
Specifications
5/Pk.
Aluminum Alloy
For use with rod up to 16mm diameter
Bosshead Clamp
Plastic headed clamping screws
Certificates
View accessory products
Product Title
Fisherbrand™ Bossheads > Includes: Plastic headed clamping screws