Learn More
Fisherbrand™ Aluminum Vial Canes
GREENER_CHOICE
Organize cryogenic samples for freezer storage or immersion in dewars. Fisherbrand™ Aluminum Vial Canes bend to fit most major brands of cryogenic vials. Available in two sizes to hold 2.0mL and 5.0mL vials.
Brand:  Fisherbrand™ HS23206
135.60 DKK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 11968084
  • 239.00 DKK / Pack of 12
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Aluminum canes hold vials for freezer storage or immersion in LN2 dewars.
  • For use in liquid and vapor phase; use approved vials only
  • Canes bend to fit all major cryogenic vial brands
  • Available in two sizes:
    • 290mm cane holds up to 5 x 2.0mL vials
    • 300mm cane holds up to 6 x 5.0mL vials
Specifications
Specifications
300 mm
6 Vials up to 5.0 mL
1 kg
Cane
For Freezer Storage or In LN2 Containers
12 Pack
Aluminum
Product Title
Fisherbrand™ Aluminum Vial Canes > Length: 300mm