Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Click to view available options
Model:
Model A2B2E2K2RD
Model A2B2P3RD
Model ABEK2P3RD
Model AXB2P3
Model HgP3
Unit Size:
Each
Description
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Specifications
Cartridge Type | Canister |
Certifications/Compliance | E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 0070-003-079-06-96-0120 Ext. 01-02-00, EC Attestation- EC Attestation, EC Attestation Number- 0070-003-079-06-96-0120 Ext. 01-02-00, Declaration |
Connection Type | RD40 |
Color | Gray, White, Green |
Material | Aluminum alloy with PE cover activated and impregnated charcoal. |
Product Type | ALU RD40 AXBP3 |
For Use With (Application) | Respiratory protection against organic gases and vapors with a boiling point below 65°C, inorganic gases and vapors, solid and liquid particles |
For Use With (Equipment) | Full-Face mask equipped with an Rd40 connector. |
Model | Model AXB2P3 |
Quantity | 1 Ea. |
Product Content Correction
Product Title
Sperian™ Honeywell™ Aluminum RD40 Filters >
Spot an opportunity for improvement?Share a Content Correction