missing translation for 'onlineSavingsMsg'
Learn More

KGW Isotherm™ Aluminum Dewar Flask

Brand:  KGW Isotherm™ 1029

Product Code. 10037750

  • 1200.00 DKK / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Spring Clips
80mm
350mm
Lid
1200 mL
Aluminum
Blue
Laboratory technique, Medicinal technique
395mm
Borosilicate Glass
67 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers