Learn More
KGW Isotherm™ Dish-Shaped Dewar Flasks
For the cooling of small samples. KGW Isotherm™ Dish-Shaped Dewar Flasks are suitable as a hot or cold bath.
Brand:  KGW Isotherm™ 1195
 View more versions of this product
Product Code. 10790972
  • 2680.00 DKK / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Easy handling
  • Suitable on magnetic stirrer
  • Full silvered
  • Made of borosilicate glass 3.3
  • Polyethylene lid (not included)
Specifications
Specifications
Lid not included
80mm
Laboratory technique, Medicinal technique
Borosilicate Glass
Aluminum
Silver
170mm
125mm
800 mL