Learn More
Welch Ilmvac™ Aluminum KF Cross Piece
Brand: Welch Ilmvac™ 710002
690.88 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Aluminum | |
1 Each | |
KF Cross Piece Adapter |
Product Content Correction
Product Title
Welch Ilmvac™ Aluminum KF Cross Piece > Size: 50 x 25mm
Spot an opportunity for improvement?Share a Content Correction