Learn More
Welch™ Aluminum KF Outer Centering Ring with Buna N O-ring
Brand:  Welch™ 701153
85.05 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10157881
  • 87.20 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Specifications
Separator 25 DN
Aluminum
1 Each
KF Outer Centering Ring
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch™ Aluminum KF Outer Centering Ring with Buna N O-ring > O.D.: 25mm