Learn More
Welch™ Aluminum KF Outer Centering Ring with Wire Mesh, Edelst. Viton O-ring
Brand: Welch™ 701195
458.10 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Separator 40 DN | |
Aluminum | |
1 Each | |
KF Outer Centering Ring |
Product Content Correction
Product Title
Welch™ Aluminum KF Outer Centering Ring with Wire Mesh, Edelst. Viton O-ring > O.D.: 40mm
Spot an opportunity for improvement?Share a Content Correction