Learn More
Welch™ Aluminum KF Outer Centering Ring
Brand:  Welch™ 701143
72.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10506292
  • 74.25 DKK / Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
Separator 25 DN
Aluminum
1 Each
KF Outer Centering Ring
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Welch™ Aluminum KF Outer Centering Ring > O.D.: 25mm