Learn More
Aluminum rod, alloy 6061, 6.35mm (0.25 in.) dia., Thermo Scientific™
220.45 DKK - 285.78 DKK
Specifications
Chemical Name or Material | Aluminum rod, Alloy 6061 |
---|---|
Name Note | 6.35mm (0.25 in.) dia. |
MDL Number | MFCD00214039 |
Odor | Odorless |
Weight | ≈0.85g/cm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389707
|
Thermo Scientific Alfa Aesar
042052.BV |
30 cm |
220.45 DKK
30cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379707
|
Thermo Scientific Alfa Aesar
042052.CE |
5 x 30 cm | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Aluminum rod, Alloy 6061 | |
MFCD00214039 | |
≈0.85g/cm |
6.35mm (0.25 in.) dia. | |
Odorless | |
Rod |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures