Learn More
Cole-Parmer™ Aluminum Scoop
Brand:  Cole-Parmer™ 07207-02
 View more versions of this product
Product Code. 11799879
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Scoop
Ribbed
6.25 in.
Aluminum
350 mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Aluminum Scoop > Capacity: 350mL