Learn More
Korff™ Aluminum Small Rolls
Brand:  Korff™ 60050
 View more versions of this product
Product Code. 12694526
  • 161.00 DKK / 10m
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Specifications
Foil Gauge: 0.030mm
Small Rolls
300mm
Aluminum
Product Title
Korff™ Aluminum Small Rolls > Width: 300mm; Foil Gauge: 0.030mm