Learn More
Heathrow Scientific™ Aluminum Test Tube Basket
Wash and store test tubes. Heathrow Scientific™ Aluminum Test Tube Basket can withstand repeated exposure to autoclaves.
Brand: Heathrow Scientific™ HS20341C
This item is not returnable.
View return policy
Description
- Constructed of lightweight aluminum
- Ideal for washing and storing test tubes
- Withstands repeated autoclaving and use in the lab
Specifications
Autoclavable | |
Aluminum | |
Washing, Autoclaving, Storing Test Tubes | |
332 x 228 x 178mm |
Product Content Correction
Product Title
Heathrow Scientific™ Aluminum Test Tube Basket >
Spot an opportunity for improvement?Share a Content Correction