Learn More
Fisherbrand™ Aluminum Weighing Boats, Pack of 100, Oil Free
Dishes for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
208.88 DKK - 671.40 DKK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Bottom Shape | Round |
Disposable | Single-use |
Material | Aluminum |
Shape | Circular |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
30182871
![]() |
Fisherbrand™
120932FRF |
5 mL |
290.94 DKK
Pack of 100 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
30182875
![]() |
Fisherbrand™
120930FRF |
70 mL |
671.40 DKK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
30182878
![]() |
Fisherbrand™
120931FRF |
25 mL |
484.90 DKK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
30182868
![]() |
Fisherbrand™
120935FRF |
42 mL |
208.88 DKK
Pack of 100 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
30182874
![]() |
Fisherbrand™
120936FRF |
80 mL |
447.60 DKK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
30182872
![]() |
Fisherbrand™
120937FRF |
140 mL |
522.20 DKK
Pack of 100 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
30182870
![]() |
Fisherbrand™
HEA14521AFRF |
20 mL |
261.10 DKK
Pack of 100 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
30182877
![]() |
Fisherbrand™
HEA14521BFRF |
34 mL |
268.56 DKK
Pack of 100 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
30182876
![]() |
Fisherbrand™
HEA14521CFRF |
50 mL |
335.70 DKK
Pack of 100 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
Description
The Aluminum Weighing Boat may be used for general-purpose weighing, dispensing, storage, food processing, evaporating or drying.
- These sturdy yet lightweight boats feature a smooth, flat bottom for sample handling.
- Oil free
Passed through a rigorous, three-step cleansing process to remove forming oil or residue for more reliable weighing and moisture sample analysis.
- Processed and packaged in clean room environment
- Lot traceable
- Volume: 70 mL
- Dimensions: 2.8 x 0.7 in. (71 x 17 mm)
- Pack of 100
- Crinkle finish
- Thickness: 0.075 mm
- Certificates for Oil-Free Aluminum Boats available upon request
Specifications
Not Autoclavable | |
Single-use | |
Circular | |
Weighing Boat |
Round | |
Aluminum | |
Non-sterile |