Learn More
Kimble Chase™ Amber Glass Ampoules Pack
Brand:  Kimble Chase™ 12040U-20
Product Code. 12737387
  • 9455.00 DKK / Pack of 1152
Estimated Shipment: 26-11-2025
Explore more special offers
Specifications
20 mL
Amber Glass
Ampule
Product Title
Kimble Chase™ Amber Glass Ampoules Pack > Capacity: 20mL