Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Use with all blue and green light chemiluminescent systems. Cytiva Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Brand: Cytiva 28-9068-39
This item is not returnable.
View return policy
Description
- Antistatic coating reduces risk of visible artifacts
- Clear base offers high contrast for easy reading of images
- Suitable for use with all blue and green light chemiluminescent systems
- Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
- Chemiluminescent detection in Western, Southern, and Northern blots
Specifications
Amersham Hyperfilm ECL | |
100 Sheets/Box | |
20.3 x 25.4 cm (8 x 10 in.) |
Chemiluminescent detection in Western, Southern, and Northern blots. | |
4°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
GEH002 hyperfilm
File size: 1.4 MB
GEH001 Western blot
File size: 2.6 MB
Product Content Correction
Product Title
Cytiva Amersham™ Hyperfilm™ ECL > Size: 8 x 10 in. (20.3 x 25.4cm); 100 sheets
Spot an opportunity for improvement?Share a Content Correction